Please login to your online account to display your discounted pricing

2-Methylfuran-3-thiolacetate, 80%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Methylfuran-3-thiolacetate
Name Note 80%
CAS 55764-25-5
CAS Min % 75.0
CAS Max % 100.0
Assay ≥75%
Assay Percent Range 80%
Molecular Formula C7H8O2S
Formula Weight 156.2
MDL Number MFCD01632595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC332670010

  SDS
thermo fisher scientific
332670010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC332670050

  SDS
thermo fisher scientific
332670050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥75%
Physical Form Liquid
Formula Weight 156.2
Quantity 1g
Color Clear colorless to light brown
TSCA TSCA
EINECS Number 259-801-0
CAS Max % 100.0
CAS Min % 75.0
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Health Hazard 1 STENCH
Infrared Spectrum Authentic
Chemical Name or Material 2-Methylfuran-3-thiolacetate
Molecular Formula C7H8O2S
MDL Number MFCD01632595
CAS 55764-25-5
Name Note 80%
Assay Percent Range 80%
Safety and Handling

STENCH