Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylformanilide, 98%

$226.01 - $789.54

Chemical Identifiers

Chemical Name or Material 2-Methylformanilide
CAS 94-69-9
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00014122
Beilstein 2205748
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2513414

  SDS
alfa aesar™
B2513414
25g Each for $226.01

AAB2513422

  SDS
alfa aesar™
B2513422
100g Each for $789.54
Description & Specifications

Specifications

Formula Weight 135.17
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 94-69-9
Chemical Name or Material 2-Methylformanilide
TSCA TSCA
Assay 98%
Quantity 25g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 287° to 289°C
Melting Point 58° to 60°C
Molecular Formula C8H9NO
Health Hazard 1 Danger
Beilstein 2205748
MDL Number MFCD00014122
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.