missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyleneglutaronitrile, TCI America™

$50.42 - $273.48

Chemical Identifiers

CAS 1572-52-7
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.128
MDL Number MFCD00001869
InChI Key NGCJVMZXRCLPRQ-UHFFFAOYSA-N
Synonym 2,4-Dicyano-1-butene
PubChem CID 74080
IUPAC Name 2-methylidenepentanedinitrile
SMILES C=C(CCC#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0222500ML View Documents TCI America
M0222500ML
500mL Each for $273.48
 
M022225ML View Documents TCI America
M022225ML
25mL Each for $50.42
 
Specifications

Chemical Identifiers

1572-52-7
106.128
NGCJVMZXRCLPRQ-UHFFFAOYSA-N
74080
C=C(CCC#N)C#N
C6H6N2
MFCD00001869
2,4-Dicyano-1-butene
2-methylidenepentanedinitrile

Specifications

98°C
500mL
C6H6N2
3276
NGCJVMZXRCLPRQ-UHFFFAOYSA-N
2-methylidenepentanedinitrile
74080
Clear Liquid at 20°C
2-Methyleneglutaronitrile
Yellow
1572-52-7
MFCD00001869
2,4-Dicyano-1-butene
C=C(CCC#N)C#N
106.128
106.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3120

TSCA : Yes