missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyldibenzo[f,h]quinoxaline, TCI America™

$117.64 - $388.06

Chemical Identifiers

CAS 536753-86-3
Molecular Formula C17H12N2
Molecular Weight (g/mol) 244.297
MDL Number MFCD16038624
InChI Key DCPGBPKLXYETTA-UHFFFAOYSA-N
PubChem CID 20720413
IUPAC Name 3-methylphenanthro[9,10-b]pyrazine
SMILES CC1=CN=C2C3=CC=CC=C3C4=CC=CC=C4C2=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M3005200MG View Documents TCI America
M3005200MG
200mg Each for $117.64
 
M30051G View Documents TCI America
M30051G
1g Each for $388.06
 
Specifications

Chemical Identifiers

536753-86-3
244.297
DCPGBPKLXYETTA-UHFFFAOYSA-N
3-methylphenanthro[9,10-b]pyrazine
C17H12N2
MFCD16038624
20720413
CC1=CN=C2C3=CC=CC=C3C4=CC=CC=C4C2=N1

Specifications

White-Yellow
200mg
C17H12N2
DCPGBPKLXYETTA-UHFFFAOYSA-N
3-methylphenanthro[9,10-b]pyrazine
20720413
Crystal-Powder at 20°C
2-Methyldibenzo[f,h]quinoxaline
130°C
536753-86-3
MFCD16038624
CC1=CN=C2C3=CC=CC=C3C4=CC=CC=C4C2=N1
244.297
244.30
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No