Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylcyclopropanemethanol, cis + trans, 98%

Chemical Identifiers

Chemical Name or Material 2-Methylcyclopropanemethanol
CAS 6077-72-1
Name Note Cis + Trans
Molecular Formula C5H10O
Formula Weight 86.13
MDL Number MFCD00009683
UN Number UN1987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2733703

  SDS
alfa aesar™
H2733703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2733709

  SDS
alfa aesar™
H2733709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.13
Quantity 1g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Assay 98%
Name Note Cis + Trans
Flash Point 39°C (102°F)
Chemical Name or Material 2-Methylcyclopropanemethanol
Molecular Formula C5H10O
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
UN Number UN1987
Odor Alcohol-like
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 133°C
MDL Number MFCD00009683
Health Hazard 1 Warning
CAS 6077-72-1

Applications
It is employed in the radical clock investigation of microsomal cytochrome P-450 hydroxylation of hydrocarbons.

Solubility
Slightly soluble in water.

Notes
Store away from halogens and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away and protect from electrostatic charges.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.