missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclopropanemethanol, 98%, mixture of cis and trans, ACROS Organics™

Chemical Identifiers

CAS 6077-72-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C5H10O
Linear Formula CH3C3H4CH2OH
Formula Weight 86.13
MDL Number MFCD00009683
Chemical Name or Material 2-Methylcyclopropanemethanol, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC214940010 SDS Thermo Fisher Scientific
214940010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6077-72-1
100.0
C5H10O
86.13
2-Methylcyclopropanemethanol, 98%
97.5
98%
CH3C3H4CH2OH
MFCD00009683
≥97.5%

Specifications

0.8700g/mL
1g
97.5
Authentic
1.4280 to 1.4330 (20°C, 589nm)
98%
CH3C3H4CH2OH
MFCD00009683
≥97.5%
Clear colorless
6077-72-1
100.0
Glass bottle
Liquid
C5H10O
86.13
2-Methylcyclopropanemethanol, 98%

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA