missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopentane-1,3-dione, 98%

CAS: 765-69-5 | C6H8O2 | 112.128 g/mol

Supplier:  Thermo Scientific Chemicals A1393614

 View more versions of this product

Catalog No. AAA1393614


Only null left
Explore available promotions

Description

Description

2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

Solubility
Solubility in hot Methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Methylcyclopentane-1,3-dione
212°C to 216°C
MFCD00001406
1237255
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methylcyclopentane-1,3-dione
13005
98%
765-69-5
C6H8O2
25 g
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
CC1C(=O)CCC1=O
112.128
112.13

Chemical Identifiers

765-69-5
112.128
HXZILEQYFQYQCE-UHFFFAOYSA-N
13005
CC1C(=O)CCC1=O
C6H8O2
MFCD00001406
2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
2-methylcyclopentane-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 212-153-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only