missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylcyclohexylamine, cis + trans, 97%

Manufacturer:  Alfa Aesar™ L0228518

 View more versions of this product

Catalog No. AAL0228518



Description

Description

Applications
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
Specifications

Specifications

2-Methylcyclohexylamine
21°C (69°F)
Air Sensitive
50g
7003-32-9
MFCD00001492
2689073
Slightly soluble in water.
CC1CCCCC1N
113.204
113.2
0.858
1.4565
149°C to 150°C
Cis + Trans
C7H15N
UN2733
2-methyl-cyclohexylamine, 2-methylcyclohexanamine, 2-methylcyclohexyl amine, 2-methylcyclohexylamine, 2-methylcyclohexylamine,c&t, 2methylcyclohexylamine, cyclohexanamine, 2-methyl, cyclohexylamine, 2-methyl, hexahydro-o-toluidine, o-methylcyclohexylamine
FEUISMYEFPANSS-UHFFFAOYSA-N
2-methylcyclohexan-1-amine
23432
97%

Chemical Identifiers

7003-32-9
113.204
FEUISMYEFPANSS-UHFFFAOYSA-N
23432
CC1CCCCC1N
C7H15N
MFCD00001492
2-methyl-cyclohexylamine, 2-methylcyclohexanamine, 2-methylcyclohexyl amine, 2-methylcyclohexylamine, 2-methylcyclohexylamine,c&t, 2methylcyclohexylamine, cyclohexanamine, 2-methyl, cyclohexylamine, 2-methyl, hexahydro-o-toluidine, o-methylcyclohexylamine
2-methylcyclohexan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 230-277-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents