missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclohexanone, TCI America™

Manufacturer:  TCI America M0197500ML

 View more versions of this product

Catalog No. M0197500ML



Specifications

Specifications

2-Methylcyclohexanone
Colorless
583-60-8
MFCD00001635
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
CC1CCCCC1=O
112.172
112.17
≥96.0% (GC)
165°C
500mL
C7H12O
2297
LFSAPCRASZRSKS-UHFFFAOYSA-N
2-methylcyclohexan-1-one
11419
Clear Liquid at 20°C

Chemical Identifiers

583-60-8
112.172
LFSAPCRASZRSKS-UHFFFAOYSA-N
11419
CC1CCCCC1=O
C7H12O
MFCD00001635
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
2-methylcyclohexan-1-one
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2411

RTECSNumber : GW1750000

TSCA : Yes

Documents