missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylcyclohexanone 96.0+%, TCI America™
Supplier: TCI America M019725ML
Specifications
2-Methylcyclohexanone | |
Colorless | |
C7H12O | |
25 mL | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
CC1CCCCC1=O | |
112.172 | |
112.17 | |
Liquid |
583-60-8 | |
165°C | |
MFCD00001635 | |
2297 | |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
2-methylcyclohexan-1-one | |
11419 | |
≥96.0% (GC) |
Chemical Identifiers
583-60-8 | |
112.172 | |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
11419 | |
CC1CCCCC1=O |
C7H12O | |
MFCD00001635 | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
2-methylcyclohexan-1-one |
Safety and Handling
EINECSNumber : (3)-2411
RTECSNumber : GW1750000
TSCA : Yes