missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126612500

 View more versions of this product

Catalog No. AC126612500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylcyclohexanone
583-60-8
100.0
Colorless to Yellow
162.0°C to 163.0°C
Authentic
Glass bottle
1.4470 to 1.4490
250 g
0.924
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CC1CCCCC1=O
112.17
112.17
Liquid
98%
97.5
-14.0°C
0.9240g/mL
48°C
97.5% min. (GC)
C7H12O
CH3C6H9(=O)
07, 14
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
LFSAPCRASZRSKS-UHFFFAOYSA-N
2-methylcyclohexan-1-one
11419
98%

Chemical Identifiers

583-60-8
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
2-methylcyclohexan-1-one
LFSAPCRASZRSKS-UHFFFAOYSA-N
11419
CC1CCCCC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-513-6

RTECSNumber : GW1750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only