Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylcinnamic acid, predominantly trans, 98%

$39.03 - $640.38

Chemical Identifiers

Chemical Name or Material 2-Methylcinnamic acid
CAS 2373-76-4
Name Note predominantly trans
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00016841
Beilstein 2500607
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1464403

  SDS
alfa aesar™
A1464403
1g Each for $39.03

AAA1464406

  SDS
alfa aesar™
A1464406
5g Each for $152.19

AAA1464414

  SDS
alfa aesar™
A1464414
25g Each for $640.38
Description & Specifications

Specifications

Formula Weight 162.19
Molecular Formula C10H10O2
CAS 2373-76-4
Chemical Name or Material 2-Methylcinnamic acid
Name Note predominantly trans
Assay 98%
MDL Number MFCD00016841
Beilstein 2500607
Health Hazard 1 Warning
Melting Point 173° to 177°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.