missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylcinnamic acid, predominantly trans, 98%

Manufacturer:  Alfa Aesar™ A1464403

 View more versions of this product

Catalog No. AAA1464403



Specifications

Specifications

2-Methylcinnamic acid
1g
2373-76-4
MFCD00016841
2-methylcinnamic acid, 3-o-tolyl acrylic acid, o-methylcinnamic acid, 2-methylcinnamic acid, predominantly trans, 2e-3-2-methylphenyl acrylic acid, e-3-2-methylphenyl-2-propenoic acid, 2e-3-2-methylphenyl prop-2-enoic acid, e-3-o-tolyl acrylic acid, e-3-o-tolylacrylic acid, 3-o-tolyl-acrylic acid
CC1=CC=CC=C1C=CC(=O)O
162.188
162.19
173°C to 177°C
predominantly trans
C10H10O2
2500607
RSWBWHPZXKLUEX-VOTSOKGWSA-N
(E)-3-(2-methylphenyl)prop-2-enoic acid
819020
98%

Chemical Identifiers

2373-76-4
162.188
RSWBWHPZXKLUEX-VOTSOKGWSA-N
819020
CC1=CC=CC=C1C=CC(=O)O
C10H10O2
MFCD00016841
2-methylcinnamic acid, 3-o-tolyl acrylic acid, o-methylcinnamic acid, 2-methylcinnamic acid, predominantly trans, 2e-3-2-methylphenyl acrylic acid, e-3-2-methylphenyl-2-propenoic acid, 2e-3-2-methylphenyl prop-2-enoic acid, e-3-o-tolyl acrylic acid, e-3-o-tolylacrylic acid, 3-o-tolyl-acrylic acid
(E)-3-(2-methylphenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents