missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific™
$454.58 - $454.58
Chemical Identifiers
CAS | 945863-21-8 |
---|---|
Molecular Formula | C13H19BN2O3 |
Molecular Weight (g/mol) | 262.116 |
MDL Number | MFCD08458483 |
InChI Key | LUMBLRWRRONQMO-UHFFFAOYSA-N |
Synonym | n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 2-n-methylaminocarbonyl-5-pyridineboronic acid pincol ester, 2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester, n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n-methylaminocarbonyl pyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine-2-carboxylic acid methylamide, 2-n-methylamidocarboxy-5-pyridine boronic acid pincol ester, 2-n-methylamidocarboxy-5-pyridineboronic acid pinacol ester, 2-n-methylaminocarbonyl pyridine-5-boronic acid pincol ester, 2-n-methylaminecarbonyl-5-pyridineboronic acid pinacol ester |
PubChem CID | 44119560 |
IUPAC Name | N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5408403
|
Thermo Scientific Chemicals
H5408403 |
1 g |
Each for $454.58
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Chemical Identifiers
945863-21-8 | |
262.116 | |
LUMBLRWRRONQMO-UHFFFAOYSA-N | |
44119560 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC |
C13H19BN2O3 | |
MFCD08458483 | |
n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 2-n-methylaminocarbonyl-5-pyridineboronic acid pincol ester, 2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester, n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n-methylaminocarbonyl pyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine-2-carboxylic acid methylamide, 2-n-methylamidocarboxy-5-pyridine boronic acid pincol ester, 2-n-methylamidocarboxy-5-pyridineboronic acid pinacol ester, 2-n-methylaminocarbonyl pyridine-5-boronic acid pincol ester, 2-n-methylaminecarbonyl-5-pyridineboronic acid pinacol ester | |
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide |
Specifications
945863-21-8 | |
C13H19BN2O3 | |
1 g | |
Slightly soluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC | |
262.116 | |
262.11 | |
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester |
113°C to 115°C | |
MFCD08458483 | |
n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 2-n-methylaminocarbonyl-5-pyridineboronic acid pincol ester, 2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester, n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n-methylaminocarbonyl pyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine-2-carboxylic acid methylamide, 2-n-methylamidocarboxy-5-pyridine boronic acid pincol ester, 2-n-methylamidocarboxy-5-pyridineboronic acid pinacol ester, 2-n-methylaminocarbonyl pyridine-5-boronic acid pincol ester, 2-n-methylaminecarbonyl-5-pyridineboronic acid pinacol ester | |
LUMBLRWRRONQMO-UHFFFAOYSA-N | |
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide | |
44119560 | |
96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen