missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54084MD

 View more versions of this product

Catalog No. AAH54084MD



Description

Description

Applications
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester
250mg
C13H19BN2O3
2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester
LUMBLRWRRONQMO-UHFFFAOYSA-N
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
44119560
96%
113°C to 115°C
945863-21-8
MFCD08458483
Slightly soluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC
262.116
262.11

Chemical Identifiers

945863-21-8
262.116
LUMBLRWRRONQMO-UHFFFAOYSA-N
44119560
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NC
C13H19BN2O3
MFCD08458483
2-methylcarbamoyl pyridine-5-boronic acid, pinacol ester
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents