Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylcarbamoyl)benzeneboronic acid, 97%

$193.95 - $193.95

Chemical Identifiers

Chemical Name or Material 2-(Methylcarbamoyl)benzeneboronic acid
CAS 874459-85-5
Molecular Formula C8H10BNO3
Formula Weight 179.08
MDL Number MFCD22190724
Synonym 2-(Methylcarbamoyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5462803

 
alfa aesar™
H5462803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH54628MD

 
alfa aesar™
H54628MD
250mg Each for $193.95
Description & Specifications

Specifications

Formula Weight 179.08
Physical Form Solid
Chemical Name or Material 2-(Methylcarbamoyl)benzeneboronic acid
Assay 97%
MDL Number MFCD22190724
CAS 874459-85-5
Synonym 2-(Methylcarbamoyl)phenylboronic acid
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H10BNO3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.