Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylbutyraldehyde, 98%

Chemical Identifiers

Chemical Name or Material 2-Methylbutyraldehyde
CAS 96-17-3
Formula Weight 86.13g/mol
MDL Number MFCD00006984
UN Number UN1989
Synonym 2-Methylbutanal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3185514

 
alfa aesar™
31855-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3185536

 
alfa aesar™
31855-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.8030g/mL
Refractive Index 1.3919
Boiling Point 90° to 92°C
Flash Point 4° (39°F)
Quantity 25g
Chemical Name or Material 2-Methylbutyraldehyde
CAS 96-17-3
Physical Form Liquid
Assay 98%
Formula Weight 86.13g/mol
MDL Number MFCD00006984
UN Number UN1989
Synonym 2-Methylbutanal
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.