missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbutylamine (contains 3-Methylbutylamine), TCI America™

Manufacturer:  TCI America M076725ML

 View more versions of this product

Catalog No. M076725ML



Specifications

Specifications

2-Methylbutylamine (contains 3-Methylbutylamine)
25mL
C5H13N
2733
VJROPLWGFCORRM-UHFFFAOYSA-N
2-methylbutan-1-amine
7283
Clear Liquid at 20°C
Yellow
96-15-1
MFCD00008147
2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
CCC(C)CN
87.166
87.17
≥85.0% (GC)

Chemical Identifiers

96-15-1
87.166
VJROPLWGFCORRM-UHFFFAOYSA-N
7283
CCC(C)CN
C5H13N
MFCD00008147
2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
2-methylbutan-1-amine
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0133

TSCA : Yes

Documents