missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-,Methylbutane, EMD Millipore

$346.37 - $346.37

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Linear Formula (CH3)2CHCH2CH3
Formula Weight 72.15
Synonym Isopentane
Chemical Name or Material 2-, Methylbutane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MMX07601 Emd Millipore Corporation
MX07601
Metal Can 4L Each for $346.37
Specifications Description & Specifications

Chemical Identifiers

78-78-4
(CH3)2CHCH2CH3
Isopentane
C5H12
72.15
2-, Methylbutane

Specifications

78-78-4
1
420°C
C5H12
800 hPa (20°C)
Isopentane
28°C (1013 hPa)
4L
0.62g/cm3 (20°C)
1.3 to 7.6%(V)
Metal Can
(CH3)2CHCH2CH3
72.15
−16°C
−57°C
2-, Methylbutane

Safety and Handling

Recommended Storage : Store below +15°C