Learn More
2-Methylbutane, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 427730010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLCSpecifications
2-Methylbutane | |
78-78-4 | |
-160.0°C | |
30.0°C | |
Glass bottle | |
1.3530 to 1.3550 | |
MFCD00009338 | |
0.0003% max. | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
72.15 | |
HPLC |
For HPLC | |
0.01 max. 1 cm cell vs HPLC−grade water at 240nm, 0.01 max. 1 cm cell vs HPLC−grade water at 400nm, 0.07 max. 1 cm cell vs HPLC−grade water at 220nm, 0.3 max. 1 cm cell vs HPLC−grade water at 210nm, 1 max. 1 cm cell vs HPLC grade water at 195nm | |
0.6200g/mL | |
−51°C | |
C5H12 | |
C2H5CH(CH3)2 | |
1 L | |
0.62 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol | |
CCC(C)C | |
72.15 | |
CHEBI:30362 | |
≥99.0% | |
Liquid |
Chemical Identifiers
78-78-4 | |
72.15 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
6556 | |
2-methylbutane |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CHEBI:30362 | |
CCC(C)C |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin drynes
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flam
GHS Signal Word: Danger
EINECSNumber : 201-142-8
RUO – Research Use Only