missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbutane, 99+%, for spectroscopy, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC167840010

 View more versions of this product

Catalog No. AC167840010



Description

Description

For UV/VIS spectroscopy
Specifications

Specifications

(1 cm cell vs water)
2-Methylbutane
−51°C
1.3530 to 1.3550
0.62
30.0°C
-160.0°C
99+%
99.0
99% min. (GC)
C2H5CH(CH3)2
01, 134
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol
CCC(C)C
72.15
CHEBI:30362
Liquid
Spectroscopy
0.0005 meq/g max.
0.6200g/mL
Glass bottle
0.0003% max.
0.02% max.
Undesignated
1L
78-78-4
100.0
C5H12
MFCD00009338
1,1,2-trimethylethane, 1,1-dimethylpropane, butane, 2-methyl, butanes, dimethylethylmethane, ethyldimethylmethane, iso-c5h12, iso-pentane, isoamylhydride, isopentane
QWTDNUCVQCZILF-UHFFFAOYSA-N
2-methylbutane
6556
72.15
99+%

Chemical Identifiers

78-78-4
72.15
QWTDNUCVQCZILF-UHFFFAOYSA-N
6556
2-methylbutane
C5H12
MFCD00009338
1,1,2-trimethylethane, 1,1-dimethylpropane, butane, 2-methyl, butanes, dimethylethylmethane, ethyldimethylmethane, iso-c5h12, iso-pentane, isoamylhydride, isopentane
CHEBI:30362
CCC(C)C
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin drynes

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flam

GHS Signal Word: Danger

EINECSNumber : 201-142-8

RTECSNumber : EK4430000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only