Learn More
2-Methylbutane, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
$164.23 - $275.45
Chemical Identifiers
CAS | 78-78-4 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009338 |
InChI Key | QWTDNUCVQCZILF-UHFFFAOYSA-N |
Synonym | isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 |
PubChem CID | 6556 |
ChEBI | CHEBI:30362 |
IUPAC Name | 2-methylbutane |
SMILES | CCC(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397221000
|
Thermo Scientific Chemicals
397221000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $164.23
|
|
||||
AC397220010
|
Thermo Scientific Chemicals
397220010 |
1 L | AcroSeal™ Glass Bottle |
Each for $275.45
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-78-4 | |
72.15 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
6556 | |
2-methylbutane |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CHEBI:30362 | |
CCC(C)C |
Specifications
78-78-4 | |
100.0 | |
Colorless | |
30.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.3530 to 1.3550 | |
MFCD00009338 | |
0.001% max. | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
72.15 | |
Extra Dry | |
2-Methylbutane, Extra Dry |
99.0 | |
-160.0°C | |
0.6200g/mL | |
−51°C | |
99% min. (GC) | |
C5H12 | |
C2H5CH(CH3)2 | |
100 mL | |
0.62 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol | |
CCC(C)C | |
72.15 | |
CHEBI:30362 | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin drynes
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flam
GHS Signal Word: Danger
EINECSNumber : 201-142-8
RUO – Research Use Only