Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Methylbiphenyl-2-carboxaldehyde, 98%

$90.50 - $326.32

Chemical Identifiers

Chemical Name or Material 2'-Methylbiphenyl-2-carboxaldehyde
CAS 7111-68-4
Molecular Formula C14H12O
Formula Weight 196.25
MDL Number MFCD03424630
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5028503

 
alfa aesar™
H5028503
1g Each for $326.32

AAH50285MD

 
alfa aesar™
H50285MD
250mg Each for $90.50
Description & Specifications

Specifications

Formula Weight 196.25
Chemical Name or Material 2'-Methylbiphenyl-2-carboxaldehyde
Assay 98%
Molecular Formula C14H12O
MDL Number MFCD03424630
CAS 7111-68-4
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.