missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbenzylamine 98.0+%, TCI America™

Manufacturer:  TCI America M094225ML

 View more versions of this product

Catalog No. M094225ML



Specifications

Specifications

2-Methylbenzylamine
Yellow
89-93-0
MFCD00008112
2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
CC1=CC=CC=C1CN
121.183
121.18
≥98.0% (GC,T)
82°C
25mL
C8H11N
2735
CJAAPVQEZPAQNI-UHFFFAOYSA-N
(2-methylphenyl)methanamine
6993
Clear Liquid at 20°C

Chemical Identifiers

89-93-0
121.183
CJAAPVQEZPAQNI-UHFFFAOYSA-N
6993
CC1=CC=CC=C1CN
C8H11N
MFCD00008112
2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
(2-methylphenyl)methanamine
Safety and Handling

Safety and Handling

RTECSNumber : DP5800000

TSCA : Yes

Documents