Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Methylbenzyl)triphenylphosphonium chloride, 98+%

$32.09 - $521.34

Chemical Identifiers

Chemical Name or Material (2-Methylbenzyl)triphenylphosphonium chloride
CAS 63368-36-5
Molecular Formula C26H24ClP
Formula Weight 402.91
MDL Number MFCD00031577
Beilstein 4172676
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1071909

  SDS
alfa aesar™
A1071909
10g Each for $32.09

AAA1071918

  SDS
alfa aesar™
A1071918
50g Each for $121.91

AAA1071930

  SDS
alfa aesar™
A1071930
250g Each for $521.34
Description & Specifications

Specifications

Formula Weight 402.91
Beilstein 4172676
TSCA TSCA
Health Hazard 1 Warning
Melting Point 270° to 275°C
Assay ≥98%
Molecular Formula C26H24ClP
CAS 63368-36-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (2-Methylbenzyl)triphenylphosphonium chloride
MDL Number MFCD00031577
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.