missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbenzyl mercaptan, 97%, Thermo Scientific Chemicals

$436.33 - $436.33

Chemical Identifiers

CAS 7341-24-4
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00022074
InChI Key PJUDFYDAJBQPEA-UHFFFAOYSA-N
Synonym 2-methylphenyl methanethiol, 2-methylbenzyl mercaptan, 2-methylbenzylmercaptan, 2-methylbenzenemethanethiol, o-methylbenzyl mercaptan, o-tolylmethanethiol, o-tolyl-methanethiol, 2-methylphenyl methane-1-thiol
PubChem CID 288028
IUPAC Name (2-methylphenyl)methanethiol
SMILES CC1=CC=CC=C1CS
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1581614
View Documents Promotion Details
Thermo Scientific Chemicals
A1581614
25 g
Each for $436.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7341-24-4
138.23
PJUDFYDAJBQPEA-UHFFFAOYSA-N
288028
CC1=CC=CC=C1CS
C8H10S
MFCD00022074
2-methylphenyl methanethiol, 2-methylbenzyl mercaptan, 2-methylbenzylmercaptan, 2-methylbenzenemethanethiol, o-methylbenzyl mercaptan, o-tolylmethanethiol, o-tolyl-methanethiol, 2-methylphenyl methane-1-thiol
(2-methylphenyl)methanethiol

Specifications

7341-24-4
Stench
1.572
25 g
2040956
2-methylphenyl methanethiol, 2-methylbenzyl mercaptan, 2-methylbenzylmercaptan, 2-methylbenzenemethanethiol, o-methylbenzyl mercaptan, o-tolylmethanethiol, o-tolyl-methanethiol, 2-methylphenyl methane-1-thiol
CC1=CC=CC=C1CS
138.23
138.23
2-Methylbenzyl mercaptan
56°C to 58°C (5 mmHg)
C8H10S
MFCD00022074
UN2810
Air Sensitive
PJUDFYDAJBQPEA-UHFFFAOYSA-N
(2-methylphenyl)methanethiol
288028
97%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only