Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylbenzyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 2-Methylbenzyl chloride
CAS 552-45-4
Molecular Formula C8H9Cl
Formula Weight 140.61
MDL Number MFCD00000899
UN Number UN3265
Beilstein 386016
Merck Index 14,10091
Synonym alpha-Chloro-o-xylene; o-Xylyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1344814

 
alfa aesar™
A1344814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1344836

 
alfa aesar™
A1344836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.61
Chemical Name or Material 2-Methylbenzyl chloride
TSCA TSCA
Assay ≥98%
Molecular Formula C8H9Cl
MDL Number MFCD00000899
CAS 552-45-4
UN Number UN3265
Beilstein 386016
Merck Index 14,10091
Synonym alpha-Chloro-o-xylene; o-Xylyl chloride
Health Hazard 1 Danger
Melting Point 2°C
Boiling Point 197° to 199°C
Quantity 25g
Flash Point 73°C (163°F)
Odor Pungent
Health Hazard 2 GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

2° to 8°C