missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbenzyl alcohol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 182630500

 View more versions of this product

Catalog No. AC182630500


 

Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Low Melting Solid
White
100.0
Authentic
98%
CH3C6H4CH2OH
2-Methylbenzyl alcohol
122.17
50g
34° to 38°C
97.5
MFCD00004622
98%
89-95-2
C8H10O

Chemical Identifiers

89-95-2
100.0
C8H10O
122.17
2-Methylbenzyl alcohol, 98%
97.5
98%
CH3C6H4CH2OH
MFCD00004622
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-954-2

TSCA : TSCA