Please login to your online account to display your discounted pricing

2-Methylbenzyl alcohol, 98%, ACROS Organics™

$147.53 - $147.53

Chemical Identifiers

Chemical Name or Material 2-Methylbenzyl alcohol
Name Note 98%
CAS 89-95-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10O
Linear Formula CH3C6H4CH2OH
Formula Weight 122.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC182630500

  SDS
thermo fisher scientific
182630500
50g Each for $147.53
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 122.17
Quantity 50g
Color White
EINECS Number 201-954-2
TSCA TSCA
Melting Point 34° to 38°C
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Linear Formula CH3C6H4CH2OH
MDL Number MFCD00004622
CAS 89-95-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methylbenzyl alcohol
Molecular Formula C8H10O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.