missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbenzoxazole 99.0+%, TCI America™

Manufacturer:  TCI America M0557500G

 View more versions of this product

Catalog No. M0557500G



Specifications

Specifications

2-Methylbenzoxazole
Undesignated
95-21-6
MFCD00005771
DQSHFKPKFISSNM-UHFFFAOYSA-N
2-methyl-1,3-benzoxazole
7225
133.15
≥99.0% (GC)
201°C
500g
C8H7NO
2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti
CC1=NC2=CC=CC=C2O1
133.15
CHEBI:51603
Clear Liquid at 20°C

Chemical Identifiers

95-21-6
133.15
DQSHFKPKFISSNM-UHFFFAOYSA-N
7225
2-methyl-1,3-benzoxazole
C8H7NO
MFCD00005771
2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti
CHEBI:51603
CC1=NC2=CC=CC=C2O1
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0156

RTECSNumber : DM4850000

TSCA : Yes

Documents