missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 95-21-6 |
---|---|
Molecular Formula | C8H7NO |
Molecular Weight (g/mol) | 133.15 |
MDL Number | MFCD00005771 |
InChI Key | DQSHFKPKFISSNM-UHFFFAOYSA-N |
Synonym | 2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti |
PubChem CID | 7225 |
ChEBI | CHEBI:51603 |
IUPAC Name | 2-methyl-1,3-benzoxazole |
SMILES | CC1=NC2=CC=CC=C2O1 |
Chemical Identifiers
95-21-6 | |
133.15 | |
DQSHFKPKFISSNM-UHFFFAOYSA-N | |
7225 | |
2-methyl-1,3-benzoxazole |
C8H7NO | |
MFCD00005771 | |
2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti | |
CHEBI:51603 | |
CC1=NC2=CC=CC=C2O1 |
Specifications
201°C | |
25g | |
C8H7NO | |
2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti | |
CC1=NC2=CC=CC=C2O1 | |
133.15 | |
CHEBI:51603 | |
Clear Liquid at 20°C | |
2-Methylbenzoxazole |
Undesignated | |
95-21-6 | |
MFCD00005771 | |
DQSHFKPKFISSNM-UHFFFAOYSA-N | |
2-methyl-1,3-benzoxazole | |
7225 | |
133.15 | |
≥99.0% (GC) |
Safety and Handling
EINECSNumber : (5)-0156
RTECSNumber : DM4850000
TSCA : Yes