Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylbenzothiazole, 98+%

$52.88 - $605.72

Chemical Identifiers

Chemical Name or Material 2-Methylbenzothiazole
CAS 120-75-2
Molecular Formula C8H7NS
Formula Weight 149.22
MDL Number MFCD00005794
Beilstein 112427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1342714

  SDS
alfa aesar™
A1342714
25g Each for $52.88

AAA1342722

  SDS
alfa aesar™
A1342722
100g Each for $141.03

AAA1342736

  SDS
alfa aesar™
A1342736
500g Each for $605.72
Description & Specifications

Specifications

Formula Weight 149.22
Chemical Name or Material 2-Methylbenzothiazole
TSCA TSCA
Assay ≥98%
Molecular Formula C8H7NS
MDL Number MFCD00005794
CAS 120-75-2
Beilstein 112427
RTECS Number DL5600000
Flash Point 102°C (215°F)
Boiling Point 237° to 238°C
Melting Point 12° to 14°C
Quantity 25g

Applications
A reagent employed in the synthesis of polycarbocyanine and thiacyanine dyes, as well as (arylfuryl)benzothiazoles.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.