Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylbenzhydrol, 98+%

$186.84 - $688.59

Chemical Identifiers

Chemical Name or Material 2-Methylbenzhydrol
CAS 5472-13-9
Molecular Formula C14H14O
Formula Weight 198.27
MDL Number MFCD00004490
Beilstein 1952703
Synonym 2-Methyldiphenylmethanol; Phenyl o-tolyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2302122

  SDS
alfa aesar™
B2302122
100g Each for $186.84

AAB2302136

  SDS
alfa aesar™
B2302136
500g Each for $688.59
Description & Specifications

Specifications

Formula Weight 198.27
Melting Point 93° to 95°C
MDL Number MFCD00004490
Molecular Formula C14H14O
Synonym 2-Methyldiphenylmethanol; Phenyl o-tolyl carbinol
Chemical Name or Material 2-Methylbenzhydrol
Boiling Point 323°C
Beilstein 1952703
TSCA TSCA
Quantity 100g
Assay ≥98%
CAS 5472-13-9