missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2315406

 View more versions of this product

Catalog No. AAB2315406



Description

Description

2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Specifications

Specifications

2-Methylbenzeneboronic acid
∼162°C to 164°C
MFCD00093526
2935796
Slightly soluble in water.
CC1=CC=CC=C1B(O)O
135.96
135.96
16419-60-6
C7H9BO2
5 g
2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl
NSJVYHOPHZMZPN-UHFFFAOYSA-N
(2-methylphenyl)boronic acid
2733267
98%

Chemical Identifiers

16419-60-6
135.96
NSJVYHOPHZMZPN-UHFFFAOYSA-N
2733267
CC1=CC=CC=C1B(O)O
C7H9BO2
MFCD00093526
2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl
(2-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

RTECSNumber : ED7777777

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only