missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylanthraquinone, TCI America™

Manufacturer:  TCI America M0156500G

 View more versions of this product

Catalog No. M0156500G



Specifications

Specifications

2-Methylanthraquinone
175°C
84-54-8
MFCD00001235
NJWGQARXZDRHCD-UHFFFAOYSA-N
2-methylanthracene-9,10-dione
6773
222.24
≥99.0% (GC)
Yellow
500g
C15H10O2
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
222.243
CHEBI:9427
Crystal-Powder at 20°C

Chemical Identifiers

84-54-8
222.243
NJWGQARXZDRHCD-UHFFFAOYSA-N
6773
2-methylanthracene-9,10-dione
C15H10O2
MFCD00001235
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CHEBI:9427
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0687

TSCA : Yes

Documents