missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylanthraquinone, TCI America™

$64.17 - $606.55

Chemical Identifiers

CAS 84-54-8
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00001235
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
PubChem CID 6773
ChEBI CHEBI:9427
IUPAC Name 2-methylanthracene-9,10-dione
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M015625G View Documents TCI America
M015625G
25g Each for $64.17
 
M0156500G View Documents TCI America
M0156500G
500g Each for $606.55
 
Specifications

Chemical Identifiers

84-54-8
222.243
NJWGQARXZDRHCD-UHFFFAOYSA-N
6773
2-methylanthracene-9,10-dione
C15H10O2
MFCD00001235
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CHEBI:9427
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Yellow
25g
C15H10O2
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
222.243
CHEBI:9427
Crystal-Powder at 20°C
2-Methylanthraquinone
175°C
84-54-8
MFCD00001235
NJWGQARXZDRHCD-UHFFFAOYSA-N
2-methylanthracene-9,10-dione
6773
222.24
≥99.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0687

TSCA : Yes