missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylanthraquinone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1450230

 View more versions of this product

Catalog No. AAA1450230



Description

Description

2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

Solubility
Soluble in water 1.234 mg/L @ 25°C.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-Methylanthraquinone
170°C to 174°C
C15H10O2
250 g
14,6021
Soluble in water 1.234mg/L @ 25°C.
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
222.243
CHEBI:9427
97%
84-54-8
209°C (408°F)
MFCD00001235
2050523
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
NJWGQARXZDRHCD-UHFFFAOYSA-N
2-methylanthracene-9,10-dione
6773
222.24

Chemical Identifiers

84-54-8
222.243
NJWGQARXZDRHCD-UHFFFAOYSA-N
6773
2-methylanthracene-9,10-dione
C15H10O2
MFCD00001235
2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
CHEBI:9427
CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
Safety and Handling

Safety and Handling

EINECSNumber : 201-539-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only