Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylanisole, 99%

Chemical Identifiers

Chemical Name or Material 2-Methylanisole
CAS 578-58-5
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00008373
UN Number UN3271
Beilstein 1857415
Synonym o-Cresyl methyl ether; 2-Methoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1389718

 
alfa aesar™
A1389718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1389730

 
alfa aesar™
A1389730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 2-Methylanisole
TSCA TSCA
Assay 99%
Molecular Formula C8H10O
MDL Number MFCD00008373
CAS 578-58-5
UN Number UN3271
Beilstein 1857415
Synonym o-Cresyl methyl ether; 2-Methoxytoluene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Flash Point 51°C (123°F)
Boiling Point 170° to 172°C
Quantity 50g

Applications
The catalytic system of disubstituted aromatics was optimized for the 2-methylanisole reduction by a proper choice of the amine/Rh ratio which should be high enough to stabilize very small colloidal rhodium particles and low enough to avoid deactivation. The total synthesis of (±)-heliannuol D and its epimer has been completed in 9 steps and 12% overall yield from 2-methylanisole. The thermal activation of 2-methylanisole (60°C) by the Ir (III) complex TpMe2Ir (C6H5) 2 (N2)(1; TpMe2= hydrotris (3, 5-dimethylpyrazolyl) borate) yielded a mixture of hydride complexes. The total synthesis of the phenolic sesquiterpene mutisianthol has been accomplished in 12 steps from the readily available 2-methylanisole. The catalytic system was optimized for the 2-methylanisole reduction by a proper choice of the amine/Rh ratio which should be high enough to stabilize very small colloidal rhodium particles and low enough to avoid deactivation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapour.