missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylamino)pyrimidine, 95%

$109.35 - $311.51

Chemical Identifiers

CAS 931-61-3
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00234126
InChI Key BQNXHDSGGRTFNX-UHFFFAOYSA-N
Synonym 2-methylaminopyrimidine, 2-pyrimidinamine, n-methyl, n-methyl-2-pyrimidinamine, methyl-pyrimidin-2-yl-amine, 2-pyrimidinamine, n-methyl-9ci, methyl pyrimidin-2-yl amine, pyrimidine, 2-methylamino, methylaminopyrimidin, methyl-2 pyrimidinylamine, methylpyrimidin-2-ylamine
PubChem CID 265776
IUPAC Name N-methylpyrimidin-2-amine
SMILES CNC1=NC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65564MD View Documents Alfa Aesar™
H65564MD
250mg Each for $109.35
 
AAH6556403 View Documents Alfa Aesar™
H6556403
1g Each for $311.51
 
Specifications

Chemical Identifiers

931-61-3
109.132
BQNXHDSGGRTFNX-UHFFFAOYSA-N
265776
CNC1=NC=CC=N1
C5H7N3
MFCD00234126
2-methylaminopyrimidine, 2-pyrimidinamine, n-methyl, n-methyl-2-pyrimidinamine, methyl-pyrimidin-2-yl-amine, 2-pyrimidinamine, n-methyl-9ci, methyl pyrimidin-2-yl amine, pyrimidine, 2-methylamino, methylaminopyrimidin, methyl-2 pyrimidinylamine, methylpyrimidin-2-ylamine
N-methylpyrimidin-2-amine

Specifications

59°C to 64°C
931-61-3
MFCD00234126
2-methylaminopyrimidine, 2-pyrimidinamine, n-methyl, n-methyl-2-pyrimidinamine, methyl-pyrimidin-2-yl-amine, 2-pyrimidinamine, n-methyl-9ci, methyl pyrimidin-2-yl amine, pyrimidine, 2-methylamino, methylaminopyrimidin, methyl-2 pyrimidinylamine, methylpyrimidin-2-ylamine
CNC1=NC=CC=N1
109.132
109.13
95%
250mg
C5H7N3
109648
BQNXHDSGGRTFNX-UHFFFAOYSA-N
N-methylpyrimidin-2-amine
265776
Solid
2-(Methylamino)pyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning