missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylamino)pyrimidine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5468203

 View more versions of this product

Catalog No. AAH5468203



Description

Description

Applications
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester
1g
C11H18BN3O2
2-methylamino pyrimidin-5-yl boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC
235.094
235.09
122°C to 125°C
904326-88-1
MFCD11878345
QDOXNCAIXITTKA-UHFFFAOYSA-N
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
46739641
96%

Chemical Identifiers

904326-88-1
235.094
QDOXNCAIXITTKA-UHFFFAOYSA-N
46739641
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC
C11H18BN3O2
MFCD11878345
2-methylamino pyrimidin-5-yl boronic acid pinacol ester
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents