missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylamino)pyridine 98.0+%, TCI America™

Manufacturer:  TCI America M082325G

 View more versions of this product

Catalog No. M082325G



Specifications

Specifications

2-(Methylamino)pyridine
Yellow
4597-87-9
MFCD00006250
SVEUVITYHIHZQE-UHFFFAOYSA-N
N-methylpyridin-2-amine
20727
Clear Liquid at 20°C
201°C
25g
C6H8N2
2-methylamino pyridine
CNC1=CC=CC=N1
108.144
108.14
≥98.0% (GC,T)

Chemical Identifiers

4597-87-9
108.144
SVEUVITYHIHZQE-UHFFFAOYSA-N
20727
CNC1=CC=CC=N1
C6H8N2
MFCD00006250
2-methylamino pyridine
N-methylpyridin-2-amine
Safety and Handling

Safety and Handling

RTECSNumber : UT5500000

TSCA : No

Documents