missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylamino)pyridine 98.0+%, TCI America™

$159.66 - $1,295.30

Chemical Identifiers

CAS 4597-87-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006250
InChI Key SVEUVITYHIHZQE-UHFFFAOYSA-N
Synonym 2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, 2-pyridinamine, n-methyl-9ci, methylamino-2 pyridine, n-2-pyridylmethylamine, n-methyl pyridimine, n-methyl-2-pyridinamine, pyridine, 2-methylamino
PubChem CID 20727
IUPAC Name N-methylpyridin-2-amine
SMILES CNC1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M082325G View Documents Tci America
M082325G
25g Each for $159.66
 
M0823500G View Documents Tci America
M0823500G
500g Each for $1,295.30
 
Specifications

Chemical Identifiers

4597-87-9
108.144
SVEUVITYHIHZQE-UHFFFAOYSA-N
20727
CNC1=CC=CC=N1
C6H8N2
MFCD00006250
2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, 2-pyridinamine, n-methyl-9ci, methylamino-2 pyridine, n-2-pyridylmethylamine, n-methyl pyridimine, n-methyl-2-pyridinamine, pyridine, 2-methylamino
N-methylpyridin-2-amine

Specifications

201°C
25g
C6H8N2
2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, 2-pyridinamine, n-methyl-9ci, methylamino-2 pyridine, n-2-pyridylmethylamine, n-methyl pyridimine, n-methyl-2-pyridinamine, pyridine, 2-methylamino
CNC1=CC=CC=N1
108.144
108.14
≥98.0% (GC,T)
Yellow
4597-87-9
MFCD00006250
SVEUVITYHIHZQE-UHFFFAOYSA-N
N-methylpyridin-2-amine
20727
Clear Liquid at 20°C
2-(Methylamino)pyridine
Safety and Handling

Safety and Handling

RTECSNumber : UT5500000

TSCA : No