missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylamino)ethanol, 99%, ACROS Organics™

Manufacturer:  Acros Organics 158265000

 View more versions of this product

Catalog No. AC158265000


 

Specifications

Specifications

2-(Methylamino)ethanol
72°C
Colorless through Yellow
500g
98.5
Authentic
Glass bottle
99%
C3H9NO
MFCD00002839
15, 6089
OPKOKAMJFNKNAS-UHFFFAOYSA-N
2-(methylamino)ethanol
8016
75.11
0.9300g/mL
159.0°C
-3.0°C
109-83-1
100.0
1.4380 to 1.4400
0.93
99%
CH3NHCH2CH2OH
04, 276
N-Methylethanolamine
CNCCO
75.111
CHEBI:21763
Liquid

Chemical Identifiers

109-83-1
75.111
OPKOKAMJFNKNAS-UHFFFAOYSA-N
8016
C3H9NO
MFCD00002839
N-Methylethanolamine
CHEBI:21763
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents