Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylamino)ethanol, 99%

$33.46 - $33.46

Chemical Identifiers

Chemical Name or Material 2-(Methylamino)ethanol
CAS 109-83-1
Molecular Formula C5H7NO
Linear Formula CH3NH(CH2)2OH
Formula Weight 75.11
MDL Number MFCD00002839
UN Number UN2735
Beilstein 1071196
Merck Index 14,6015
Synonym N-Methylethanolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB23180AP

  SDS
alfa aesar™
B23180AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB23180AE

  SDS
alfa aesar™
B23180AE*
100mL Each for $33.46
Description & Specifications

Specifications

Formula Weight 75.11
Linear Formula CH3NH(CH2)2OH
Melting Point −5°C
Merck Index 14,6015
Boiling Point 159°C
Quantity 500mL
MDL Number MFCD00002839
Beilstein 1071196
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Flash Point 74°C (165°F)
RTECS Number KL6650000
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-(Methylamino)ethanol
CAS 109-83-1
Assay 99%
UN Number UN2735
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Odor Ammonia-like
TSCA TSCA
Synonym N-Methylethanolamine
Molecular Formula C5H7NO

Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

Solubility
Miscible with water, alcohol and ether.

Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.