missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Methylamino)butane hydrochloride, 94%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2754703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in hydrochloric acid.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.Â
Specifications
2-(Methylamino)butane hydrochloride | |
76°C to 82°C | |
MFCD07777098 | |
Hygroscopic | |
Soluble in hydrochloric acid. | |
CCC(C)NC.Cl | |
123.624 | |
123.62 |
75098-40-7 | |
C5H14ClN | |
1 g | |
n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1 | |
ICBNZSHUWSBMCL-UHFFFAOYSA-N | |
N-methylbutan-2-amine;hydrochloride | |
21273362 | |
94% |
Chemical Identifiers
75098-40-7 | |
123.624 | |
ICBNZSHUWSBMCL-UHFFFAOYSA-N | |
21273362 | |
CCC(C)NC.Cl |
C5H14ClN | |
MFCD07777098 | |
n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1 | |
N-methylbutan-2-amine;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only