missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5453403

 View more versions of this product

Catalog No. AAH5453403



Description

Description

Notes
Incompatible with oxidizing agents, air, heat. Keep at cool temperature.
Specifications

Specifications

2-Methylamino-3-methylpyridine-5-boronic acid pinacol ester
1g
C13H21BN2O2
2-methylamino-3-methylpyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)NC)C
248.133
248.13
105°C to 108°C
1111637-92-3
MFCD12923387
VZNAKHVRKWXDMU-UHFFFAOYSA-N
N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
57416512
96%

Chemical Identifiers

1111637-92-3
248.133
VZNAKHVRKWXDMU-UHFFFAOYSA-N
57416512
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)NC)C
C13H21BN2O2
MFCD12923387
2-methylamino-3-methylpyridine-5-boronic acid pinacol ester
N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents