missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Methylacetophenone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 126160050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2'-Methylacetophenone | |
577-16-2 | |
100.0 | |
214°C | |
Authentic | |
Glass bottle | |
1.5290 to 1.533 | |
MFCD00008734 | |
07, 306 | |
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl | |
CC1=CC=CC=C1C(=O)C | |
134.18 | |
134.18 |
98% | |
97.5 | |
1.0260g/mL | |
75°C | |
97.5% min. (GC) | |
C9H10O | |
CH3C6H4COCH3 | |
5g | |
1.026 | |
YXWWHNCQZBVZPV-UHFFFAOYSA-N | |
1-(2-methylphenyl)ethanone | |
11340 | |
98% |
Chemical Identifiers
577-16-2 | |
134.18 | |
YXWWHNCQZBVZPV-UHFFFAOYSA-N | |
11340 | |
CC1=CC=CC=C1C(=O)C |
C9H10O | |
MFCD00008734 | |
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl | |
1-(2-methylphenyl)ethanone |
Safety and Handling
EINECSNumber : 209-408-5
TSCA : TSCA
RUO â Research Use Only