Learn More
2'-Methylacetanilide, 98+%, Thermo Scientific Chemicals
$75.08 - $259.12
Chemical Identifiers
CAS | 120-66-1 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.193 |
MDL Number | MFCD00014961 |
InChI Key | BPEXTIMJLDWDTL-UHFFFAOYSA-N |
Synonym | o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide |
PubChem CID | 8443 |
IUPAC Name | N-(2-methylphenyl)acetamide |
SMILES | CC1=CC=CC=C1NC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1323514
|
Thermo Scientific Chemicals
A1323514 |
25 g |
Each for $75.08
|
|
|||||
AAA1323522
|
Thermo Scientific Chemicals
A1323522 |
100 g |
Each for $259.12
|
|
|||||
Description
Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates.Â
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates.Â
Solubility
It is slightly soluble in acetone, chloroform, toluene, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under ordinary conditions.
Chemical Identifiers
120-66-1 | |
149.193 | |
BPEXTIMJLDWDTL-UHFFFAOYSA-N | |
8443 | |
CC1=CC=CC=C1NC(=O)C |
C9H11NO | |
MFCD00014961 | |
o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide | |
N-(2-methylphenyl)acetamide |
Specifications
120-66-1 | |
1.16 | |
C9H11NO | |
25 g | |
14,74 | |
It is slightly soluble in acetone,chloroform,toluene,benzene. | |
CC1=CC=CC=C1NC(=O)C | |
149.193 | |
149.19 | |
2'-Methylacetanilide |
109°C to 112°C | |
296°C | |
MFCD00014961 | |
2207054 | |
o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide | |
BPEXTIMJLDWDTL-UHFFFAOYSA-N | |
N-(2-methylphenyl)acetamide | |
8443 | |
≥98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 204-414-4
RTECSNumber : AN2900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only