Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 2-Methyl-D-phenylalanine
CAS 80126-54-1
Formula Weight 179.22
MDL Number MFCD01860870
Beilstein 6175785
Synonym D-Phe(2-Me)-OH; (R)-2-Amino-3-(2-methylphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6510403

  SDS
alfa aesar™
H6510403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6510406

  SDS
alfa aesar™
H6510406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65104MD

  SDS
alfa aesar™
H65104MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material 2-Methyl-D-phenylalanine
MDL Number MFCD01860870
CAS 80126-54-1
Physical Form Powder
Beilstein 6175785
Synonym D-Phe(2-Me)-OH; (R)-2-Amino-3-(2-methylphenyl)propionic acid
Color Off-white
Quantity 1g
Assay 95%
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C