Please login to your online account to display your discounted pricing

2-Methyl-beta-naphthothiazole, 98%, ACROS Organics™

$126.63 - $126.63

Chemical Identifiers

Chemical Name or Material 2-Methyl-β-naphthothiazole
CAS 2682-45-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12 H9 N S
Formula Weight 199.27
MDL Number MFCD00004951
Synonym 2-Methylnaphtho[1, 2-d]thiazole
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC127190100

  SDS
thermo fisher scientific
127190100
Glass bottle 10g Each for $126.63
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 199.27
Quantity 10g
Color Light yellow to beige-brown
Melting Point 93° to 97°C
Beilstein 27, 69
CAS Max % 100.0
Chemical Name or Material 2-Methyl-β-naphthothiazole
MDL Number MFCD00004951
Molecular Formula C12 H9 N S
Assay Percent Range 98%
CAS Min % 97.5
CAS 2682-45-3
Packaging Glass bottle
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Synonym 2-Methylnaphtho[1, 2-d]thiazole